missing translation for 'onlineSavingsMsg'
Learn More

TCTN1 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18416272 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416272 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416272 Supplier Novus Biologicals Supplier No. NBP177182UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCTN1 Polyclonal antibody specifically detects TCTN1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTN1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias TECT1FLJ21127, tectonic 1, tectonic family member 1, tectonic-1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human TCTN1. The immunogen is located within amino acids 30 - 80 of TCTN1. Amino Acid Squence: PSTRPPGTPRAPGPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 79600
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.