missing translation for 'onlineSavingsMsg'
Learn More

TCTEX1D4 Antibody - BSA Free, Novus Biologicals™

Product Code. 18337820 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337820 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337820 Supplier Novus Biologicals Supplier No. NBP281863

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCTEX1D4 Polyclonal specifically detects TCTEX1D4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTEX1D4
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS
Gene Alias Tctex1 domain containing 4
Gene Symbols TCTEX1D4
Host Species Rabbit
Immunogen TCTEX1D4 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human TCTEX1D4. The immunogen is located within the first 50 amino acids of TCTEX1D4. Amino Acid Squence: RPLPPGRQEEENAKDSGRK
Molecular Weight of Antigen 24 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343521
Test Specificity TCTEX1D4 is predicted to not cross-react with other members of the TCTEX-type family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.