missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTEX1D2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81831
This item is not returnable.
View return policy
Description
TCTEX1D2 Polyclonal specifically detects TCTEX1D2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| TCTEX1D2 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| TCTEX1D2 | |
| TCTEX1D2 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human TCTEX1D2. The immunogen is located within the first 50 amino acids of TCTEX1D2. Amino Acid Squence: DGVPEAEKNAGEPENT | |
| Affinity Purified | |
| RUO | |
| 255758 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| MGC33212, Tctex1 domain containing 2, tctex1 domain-containing protein 2 | |
| Rabbit | |
| 16 kDa | |
| 0.1 mg | |
| Primary | |
| TCTEX1D2 is predicted to not cross-react with other members of the TCTEX-type family. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction