missing translation for 'onlineSavingsMsg'
Learn More

TCTEX1D1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18395701 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395701 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395701 Supplier Novus Biologicals Supplier No. NBP281830

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCTEX1D1 Polyclonal specifically detects TCTEX1D1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTEX1D1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS
Gene Alias FLJ40873, MGC125768, RP11-266I14.2, Tctex1 domain containing 1
Gene Symbols TCTEX1D1
Host Species Rabbit
Immunogen TCTEX1D1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human. The immunogen is located within amino acids 30 - 80 of TCTEX1D1. Amino Acid Squence: SYMEEPSQRDDISRLT
Molecular Weight of Antigen 20 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200132
Test Specificity At least three isoforms are known to exist; this antibody will detect the two largest isoforms. TCTEX1D1 is predicted to not cross-react with other members of the TCTEX-type family.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.