missing translation for 'onlineSavingsMsg'
Learn More

TCTE3 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18430747 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430747 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430747 Supplier Novus Biologicals Supplier No. NBP281956AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCTE3 Polyclonal antibody specifically detects TCTE3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTE3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias MGC142199, T-complex testis-specific protein 3, T-complex-associated testis-expressed protein 3, t-complex-associated-testis-expressed 3, tcte-3, tctex1 domain-containing protein 3, TCTEX1D3Tctex1 domain containing 3
Host Species Rabbit
Immunogen TCTE3 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human TCTE3. The immunogen is located within the first 50 amino acids of TCTE3. Amino Acid Squence: APVTPRKERRPSMFEKEAY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6991
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.