missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TCS OX2 29

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent and selective OX2 receptor antagonist (IC50 = 40 nM). Displays >250-fold selectivity for OX2 over OX1 and a range of receptors, ion channels and transporters. Inhibits orexin A induced IP3 accumulation and ERK1/2 phosphorylation in CHO cells transfected with the OX2 receptor.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 433.97 |
| Purity | >98% |
| Molecular Weight (g/mol) | 433.977 |
| Chemical Name or Material | TCS OX2 29 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction