missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCS-OX2-29, MedChemExpress
Description
TCS-OX2-29 is a potent, high affinities and selective orexin-2 receptor (OX2R) antagonist with an IC50 value of 40 nM and a pKI value of 7.5. TCS-OX2-29 displays ∼250-fold selectivity for OX2 over OX1.
Specifications
Specifications
| Chemical Name or Material | TCS-OX2-29 |
| CAS | 372523-75-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H31N3O3 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 25 mg/mL (62.89 mM; Need ultrasonic) ∣Ethanol : 25 mg/mL (62.89 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.69 mM; Need ultrasonic) |
| SMILES | CC(C)(C)[C@H](NCC1=CC=NC=C1)C(N2CC3=C(C=C(OC)C(OC)=C3)CC2)=O |
| Molecular Weight (g/mol) | 397.51 |
| Formula Weight | 397.51 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?