missing translation for 'onlineSavingsMsg'
Learn More

TCS 401, MedChemExpress

Product Code. 16477807
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16477807

Brand: Medchem Express HY12312

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TCS 401 is a selective inhibitor of protein tyrosine phosphatase 1B (PTP1B).

TCS 401 is a selective inhibitor of protein tyrosine phosphatase 1B (PTP1B).

Specifications

Chemical Name or Material TCS 401
Physical Form Solid
CAS 243966-09-8
Color Earth Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C10H11ClN2O5S
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information 0.1 M NaOH : 6.67 mg/mL (21.75 mM; ultrasonic and adjust pH to 9 with NaOH)
SMILES O=C(C1=C(NC(C(O)=O)=O)SC2=C1CCNC2)O.[H]Cl
Molecular Weight (g/mol) 306.72
Formula Weight 306.72
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.