missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TCS 359

Product Code. 18075592
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18075592 10 mg 10mg
18055182 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18075592 Supplier Tocris Bioscience™ Supplier No. 2591/10
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent FLT3 inhibitor

Potent inhibitor of FLT3 receptor tyrosine kinase (IC50 = 42 nM) that displays selectivity over a range of other kinases. Exhibits antiproliferative effects on MV4-11 cells, a human acute myelogenous leukemia cell line expressing a constitutively active mutant FLT3 (IC50 = 340 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 301305-73-7
Molecular Formula C18H20N2O4S

Specifications

Quantity 10 mg
Formula Weight 360.43
Purity >99%
Molecular Weight (g/mol) 360.428
Chemical Name or Material TCS 359
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.