missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TCS 2314

Product Code. 18044232
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18044232 50 mg 10mg
18035322 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18044232 Supplier Tocris Bioscience™ Supplier No. 3900/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

α4β1 (VLA-4) antagonist

Integrin very late antigen-4 (VLA-4; α4β1) antagonist (IC50 = 4.4 nM). Blocks the activation of inflammatory cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 317353-73-4
Molecular Formula C28H34N4O6

Specifications

Quantity 50 mg
Formula Weight 522.59
Purity 98%
Molecular Weight (g/mol) 522.602
Chemical Name or Material TCS 2314
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.