missing translation for 'onlineSavingsMsg'
Learn More

TCS 2314, MedChemExpress

Product Code. 16467807
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467807 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16467807 Supplier Medchem Express Supplier No. HY12308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TCS 2314 (compound 3) is orally active and selective very late antigen-4 (VLA-4, α4β1, CD49d/CD29) antagonist with an IC50 of 4.4 nM.

TCS 2314 (compound 3) is orally active and selective very late antigen-4 (VLA-4, α4β1, CD49d/CD29) antagonist with an IC50 of 4.4 nM.

Specifications

Chemical Name or Material TCS 2314
Physical Form Solid
CAS 317353-73-4
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C28H34N4O6
For Use With (Application) Neuroscience-Neuromodulation
SMILES O=C(O)CC1CCN(C([C@H]2N(C(CC3=CC=C(NC(NC4=CC=CC=C4C)=O)C=C3)=O)CCOC2)=O)CC1
Molecular Weight (g/mol) 522.59
Formula Weight 522.59
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.