missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCS 2312, Tocris Bioscience™

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Potent and selective checkpoint kinase 1 (chk1) inhibitor (Ki = 0.38 nM, EC50 = 60 nM). Enhances the cell killing activity of Gemcitabine (Cat. No. 3259) in breast and prostate cancer cell lines and displays antiproliferative effects in vitro.
Specifications
Specifications
| CAS | 838823-31-7 |
| Molecular Formula | C25H24N4O2 |
| Quantity | 1 mg |
| Synonym | 4'-5-3-cyclopropylamino methyl phenyl amino-1h-pyrazol-3-yl-1,1'-biphenyl-2,4-diol, 4-5-4-cyclopropylaminomethyl-phenylamino-2h-pyrazol-3-yl-biphenyl-2,4-diol, tc0 |
| InChI Key | BYFCMDWKTJJQGR-UHFFFAOYSA-N |
| SMILES | C1CC1NCC2=CC=C(C=C2)NC3=CC(=C4C=CC(=C5C=CC(=O)C=C5O)C=C4)NN3 |
| IUPAC Name | 4-[4-[5-[4-[(cyclopropylamino)methyl]anilino]-1,2-dihydropyrazol-3-ylidene]cyclohexa-2,5-dien-1-ylidene]-3-hydroxycyclohexa-2,5-dien-1-one |
| Molecular Weight (g/mol) | 412.493 |
| PubChem CID | 11258443 |
| Formula Weight | 412.48 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction