missing translation for 'onlineSavingsMsg'
Learn More

TCS 1105, TRC

Product Code. 30426611
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426611 10 mg 10mg
30423249 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426611 Supplier Toronto Research Chemicals Supplier No. TRCT12713510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 185391-33-7
Molecular Formula C17 H13 F N2 O2
Molecular Weight (g/mol) 296.296
Synonym TCS 1105, N-[(-4-Fluorophenyl)methyl]-α-oxo-1H-indole-3-acetamide
IUPAC Name N-[(4-fluorophenyl)methyl]-2-(1H-indol-3-yl)-2-oxoacetamide
SMILES Fc1ccc(CNC(=O)C(=O)c2c[nH]c3ccccc23)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H13FN2O2/c18-12-7-5-11(6-8-12)9-20-17(22)16(21)14-10-19-15-4-2-1-3-13(14)15/h1-8,10,19H,9H2,(H,20,22)
Formula Weight 296.096
Chemical Name or Material N-[(-4-Fluorophenyl)methyl]-alpha-oxo-1H-indole-3-acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.