missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TCS 1102
GREENER_CHOICE

Product Code. 18096012
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18096012 10 mg 10mg
18085802 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18096012 Supplier Tocris Bioscience™ Supplier No. 3818/10
  • ONLINE EXCLUSIVE PRICE
    £187.00
    £176.40 / 10mg
    Save £10.60
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Potent, dual orexin receptor antagonist

Potent, dual orexin receptor antagonist (Ki values are 0.2 and 3 nM for OX2 and OX1 receptors respectively). Inhibits ADL-orexin B-mediated locomotion following i.p. administration in vivo and blocks orexin-A (Cat.No.1455) mediated increases in feeding behaviour. Brain penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916141-36-1
Molecular Formula C27H26N4O2S

Specifications

Quantity 10 mg
Formula Weight 470.59
Purity >98%
Molecular Weight (g/mol) 470.591
Chemical Name or Material TCS 1102
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.