missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR1-N6 Antibody (CACTB6A), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61060
This item is not returnable.
View return policy
Description
TCR1-N6 Monoclonal specifically detects TCR1-N6 in Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| TCR1-N6 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10 - 1:1000 | |
| Mouse | |
| Unpurified | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunohistochemistry, Functional Assay | |
| CACTB6A | |
| Unconjugated | |
| Ascites with 0.09% Sodium Azide | |
| Bovine ConA stimulated mononuclear leukocytes | |
| 0.1 mg | |
| Bovine | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction