Learn More
TCR V gamma 2 Monoclonal Antibody (UC3-10A6), PE-Cyanine7, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 25-5828-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The UC3-10A6 monoclonal antibody reacts with the mouse V gamma 2 (Vg2) delta chain of the gamma delta T cell receptor (TCR). The gamma delta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gamma delta TCR-bearing T cells are not yet fully understood. Studies suggest that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulating the immune response during bacterial infection. Applications Reported: This UC3-10A6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This UC3-10A6 antibody has been tested by flow cytometric analysis of mouse splenocytes or lymph node cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.
TCR V gamma 2 Monoclonal antibody specifically detects TCR V gamma 2 in Mouse samples. It is validated for Flow Cytometry.Specifications
TCR V gamma 2 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Armenian Hamster | |
25 μg | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
UC3-10A6 | |
PE-Cyanine7 | |
TCR V gamma2; TCRV gamma 2; TCRV gamma2; TCRV y2; Vg2; Vgamma2 | |
Affinity chromatography | |
RUO | |
Mouse | |
Antibody | |
IgG |
For Research Use Only.