missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCR V delta 1 Monoclonal Antibody (TS8.2), PE-Cyanine7, eBioscience™, Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£172.00 - £356.00

Specifications

Antigen TCR V delta 1
Clone TS8.2
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16300784
View Documents
Invitrogen™
25-5679-41
25 Tests
£172.00
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16310784
GREENER_CHOICE
View Documents
Invitrogen™
25-5679-42
100 Tests
£356.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mouse Monoclonal Antibody

The T Cell Receptor gamma-delta (TCR gamma delta) is a heterodimeric membrane receptor that consists of the highly variable gamma and delta chains expressed as part of a complex with the invariant CD3 proteins. Gamma-delta T cells are less frequent than alpha-beta T cells but compose a substantial part of T cells found at epithelial borders including the gut mucosa resident intraepithelial lymphocytes (IEL). The antigenic stimuli required for gamma-delta T cell receptor activation are not well understood and are thought to be mainly MHC class-independent.
Specifications

Specifications

TCR V delta 1
5 μL/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
6964
Full length protein of Human TCR delta v1.
Primary
4°C, store in dark, DO NOT FREEZE!
TRD
TS8.2
Flow Cytometry
PE-Cyanine7
Mouse
Human
0
TRD
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.