missing translation for 'onlineSavingsMsg'
Learn More

TCR V delta 1 Antibody (TS8.2), Novus Biologicals™

Product Code. 18430452 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
50 ug
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430452 50 ug 50µg
18110012 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18430452 Supplier Novus Biologicals Supplier No. NBP22250650ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TCR V delta 1 Monoclonal specifically detects TCR V delta 1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR V delta 1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone TS8.2
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1 - 2 ug, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Gene Alias T cell receptor delta locus, T-cell antigen receptor, delta polypeptide, T-cell receptor, delta (V,D,J,C), TCRD, TCRD, TRD@, TCRDV1, TRD@
Gene Symbols TRD
Host Species Mouse
Immunogen Human TCR V delta 1
Purification Method Protein G purified
Quantity 50 ug
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6964
Test Specificity This antibody reacts with Human.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.