missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCR V beta 5.1/5.2 Monoclonal Antibody (MR9-4), eFluor™ 450, eBioscience™
GREENER_CHOICE

Mouse Monoclonal Antibody

£252.00

Specifications

Antigen TCR V beta 5.1/5.2
Clone MR9-4
Concentration 0.2 mg/mL
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Applications Flow Cytometry
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15589766
GREENER_CHOICE
View Documents
Invitrogen™
48-5796-82
100 μg
£252.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: This MR9-4 monoclonal antibody reacts with the mouse T cell receptor (TCR) V beta 5.1/5.2 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The MR9-4 antibody recognizes the V beta 5.1 and 5.2 chains on T cells from mouse strains with the b haplotype of the Tcrb gene, including C57BL/6, B10, and C58/J. V beta 5.1 and 5.2 are deleted in mouse strains that express MHC Class II I-E, including Balb/c, C3H, and DBA/2. Reports also indicate that V beta 5+ T cells recognize the OVA peptide. Applications Reported: This MR9-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MR9-4 antibody has been tested by flow cytometric analysis of mouse lymph node cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
Specifications

Specifications

TCR V beta 5.1/5.2
0.2 mg/mL
Flow Cytometry
eFluor 450
Mouse
RUO
PBS with 0.09% sodium azide; pH 7.2
Primary
Affinity chromatography
MR9-4
4°C, store in dark, DO NOT FREEZE!
Monoclonal
Liquid
IgG1 κ
Mouse
TCR V beta5.1; TCR V beta5.2; TCRV beta 5.1; TCRV beta 5.2; TCRV beta5.1; TCRV beta5.2; Vb5.1; Vb5.2; Vbeta5.1; Vbeta5.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.