Learn More
TCR V beta 10b Monoclonal Antibody (B21.5), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£75.25 - £134.00
Specifications
Antigen | TCR V beta 10b |
---|---|
Clone | B21.5 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15239859
|
Affymetrix eBioscience
13-5805-80 |
25 μg |
£75.25
25µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15249859
|
Affymetrix eBioscience
13-5805-82 |
100 μg |
£134.00
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The B21.5 monoclonal antibody reacts with the Vbeta10 T Cell Receptor (TCR) in mice of the b haplotype of the TCR Vbeta gene complex, which includes AKR, BALB/c, CBA, C3H/He, C57BL, DBA/1 and DBA/2 mice. It does not react with Vbeta10 in mice of the a or c haplotypes of the TCR Vbeta gene complex, including C57BR, C57L, SJL, and SWR mice. Applications Reported: This B21.5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B21.5 antibody has been tested by flow cytometric analysis of C57Bl/6 mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.Specifications
TCR V beta 10b | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
T cell receptor beta chain; Tcrb; TCRbeta; Tib | |
Tcrb | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Tcrb |
B21.5 | |
Flow Cytometry | |
Biotin | |
Rat | |
Mouse | |
0 | |
21577 | |
IgG2a λ | |
Affinity chromatography | |
Antibody |
For Research Use Only.