missing translation for 'onlineSavingsMsg'
Learn More

TCR V alpha 12.1 Antibody (6D6.6), Novus Biologicals™

Product Code. 18408235 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408235 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408235 Supplier Novus Biologicals Supplier No. NBP22249150ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TCR V alpha 12.1 Monoclonal specifically detects TCR V alpha 12.1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR V alpha 12.1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay
Classification Monoclonal
Clone 6D6.6
Concentration 0.15 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1 - 1:100, Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional
Gene Accession No. P01848
Gene Alias T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha cluster, TCR V alpha 12 TCRA, TCRD, TRA
Gene Symbols TRA
Host Species Mouse
Immunogen Human TCR V alpha 12.1
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6955
Test Specificity This antibody reacts with Human.
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.