missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR Vβ5.1 Mouse anti-Human , PE, Clone: LC4, BD Biosciences™
Mouse Monoclonal Antibody
£217.51 - £267.00
Specifications
| Antigen | TCR Vβ5.1 |
|---|---|
| Clone | LC4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17207824
|
BD Biosciences
567043 |
25 Tests |
£267.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17297814
|
BD Biosciences
567042 |
100 Tests |
£227.00 £217.51 / 100 tests Save £9.49 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCR Vβ5.1 Monoclonal antibody specifically detects TCR Vβ5.1 in Human samples. It is validated for Flow CytometrySpecifications
| TCR Vβ5.1 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human T acute lymphoblastic leukemia Cell Line | |
| Primary |
| LC4 | |
| Monoclonal | |
| Mouse | |
| Human | |
| TCR V beta 5.1; TCR Vb5.1; TRBV5-1; TRBV51 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title