missing translation for 'onlineSavingsMsg'
Learn More

TCR Vβ2 Rat anti-Mouse, BUV615, Clone: B20.6, BD OptiBuild™

Product Code. 17253005
Change view
Click to view available options
Quantity:
1 x 50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17253005 1 x 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17253005 Supplier BD Biosciences Supplier No. 752313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

TCR Vβ2 Monoclonal antibody specifically detects TCR Vβ2 in Mouse samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR Vβ2
Applications Flow Cytometry
Classification Monoclonal
Clone B20.6
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Ms V Bta 2 TCR BUV615 B20.6 50 μg
Host Species Rat
Immunogen Mouse Cytotoxic T-Cell Clone Soluble αβ TCR
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity TCR Vβ2
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a κ
Reactivity Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.