Learn More
TCR gamma/delta Monoclonal Antibody (UC7-13D5), Functional Grade, eBioscience™, Invitrogen™
Description
The UC7-13D5 MAb reacts with the mouse gamma delta TCR complex. The gamma delta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gamma delta TCR-bearing T cells are not yet fully understood; it is suggested that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulation of the immune response and during bacterial infection.
Specifications
Specifications
| Antigen | TCR gamma/delta |
| Applications | Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Clone | UC7-13D5 |
| Concentration | 1 mg/mL |
| Conjugate | Functional Grade |
| Formulation | TBS with no preservative; pH 8.0 |
| Gene | Tcrg |
| Gene Alias | T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@ |
| Host Species | Armenian Hamster |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.