Learn More
Invitrogen™ TCR gamma/delta Monoclonal Antibody (eBioGL3 (GL-3, GL3)), FITC, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody
£78.25 - £234.00
Specifications
| Antigen | TCR gamma/delta |
|---|---|
| Clone | eBioGL3 (GL-3, GL3), GL3) |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15288239
|
Invitrogen™
11-5711-81 |
50 μg |
£78.25
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15298239
![]() |
Invitrogen™
11-5711-82 |
100 μg |
£168.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15208249
|
Invitrogen™
11-5711-85 |
500 μg |
£234.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The GL3 monoclonal antibody (mAb) reacts with the gamma delta T-cell Receptor complex (TCR) on all gamma delta TCR bearing T cells, but it does not react with alpha beta TCR. The gamma delta TCR is present on cells in the thymus, epidermis, epithelial lining of the intestine, peritoneal cavity, and lymphoid tissues. Applications Reported: This GL3 (GL-3) antibody has been reported for use in flow cytometric analysis. It is recommended to prestain cells with anti-mouse CD16/32 (cat. 14-0161) to prevent non-specific Fc-mediated binding of the GL3 monoclonal antibody. Applications Tested: This eBioGL3 (GL-3, GL3) antibody has been tested by flow cytometric analysis of mouse lymph node cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
TCR gamma/delta (T-cell receptor gamma/delta) are specialized T-cells in the immune system. The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory auto-destructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). However, a small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.Specifications
| TCR gamma/delta | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| RGD1311300; similar to T cell receptor V delta 6; T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@ | |
| Tcrd, Tcrg | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBioGL3 (GL-3, GL3), GL3) | |
| Flow Cytometry | |
| FITC | |
| Armenian Hamster | |
| Mouse | |
| 0 | |
| 110066, 110067 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.