missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR gamma/delta Antibody (V65) - Low Endotoxin and Azide Free, Novus Biologicals™
Description
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Rat samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay
Specifications
Specifications
| Antigen | TCR gamma/delta |
| Applications | Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay |
| Classification | Monoclonal |
| Clone | V65 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1-4 ug/mL, Immunoprecipitation, Immunohistochemistry-Frozen, Functional |
| Formulation | Low endotoxin azide free phosphate buffered saline (PBS) solution, 0.2 um filter sterilized. Endotoxin level is less than 0.01 EU/ug of the protein, as determined by the LAL test. |
| Gene Alias | T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?