missing translation for 'onlineSavingsMsg'
Learn More

TCR DO11.10 Monoclonal Antibody (KJ1-26 (KJ126, KJI-26, KJI26)), Biotin, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5808-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15279859

  • £82.00 / 50µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The KJ1-26 anti-idiotypic monoclonal antibody binds the T-cell receptor (TCR) expressed on DO11.10 and DO11.10.24 murine T cell hybridomas, but not on any other T cell hybridomas tested.The TCR expressed on DO11.10 recognizes an ovalbumin peptide in the context of I-A^{d^} major histocompatibility molecules.The use of DO11.10 and KJ1-26 have proven to be instrumental in describing many of the intricate details of T cell immunity. Applications Reported: This KJ1-26 (KJ126, KJI-26, KJI26) antibody has been reported for use in flow cytometric analysis. Applications Tested: This KJ1-26 (KJ126, KJI-26, KJI26) antibody has been tested by flow cytometric analysis of the DO11.10 T cell hybridoma. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

TCR DO11 (DO11.10) is a receptor found on lymphocytes of DO11.10 transgenic mice and T-cell receptors of BALB/c-derived DO11.10 and DO11.10.24 T cell hybridomas. The DO11.10 TCR recognizes a C-terminal ovalbumin peptide (OVA 323-339) in the context of I-Ad major histocompatibility molecules. The use of antibodies against DO11.10 have proven to be instrumental in describing many of the intricate details of T cell immunity, including T cell activation, development, and function.
Specifications

Specifications

TCR DO11.10
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P01849
Tcra
Affinity chromatography
RUO
21473
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
KJ1-26 (KJ126, KJI-26, KJI26), KJI-26, KJI26)
Biotin
Tcra
D011.10; T cell receptor alpha chain; Tcra; Tcralpha
Mouse
50 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.