missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP11L2 Polyclonal Antibody

Brand: Thermo Scientific PA5113246
This item is not returnable.
View return policy
Description
Specifications
| TCP11L2 | |
| Polyclonal | |
| Unconjugated | |
| TCP11L2 | |
| t-complex 11 like 2; t-complex 11, testis-specific-like 2; T-complex protein 11-like protein 2; TCP11L2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 255394 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8N4U5 | |
| TCP11L2 | |
| Recombinant Human T-complex protein 11-like protein 2 protein (1-210AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction