missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP11L2 Antibody (OTI2A10), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74477PCP
This item is not returnable.
View return policy
Description
TCP11L2 Monoclonal specifically detects TCP11L2 in Human samples. It is validated for Flow Cytometry.
Specifications
| TCP11L2 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| TCP11L2 | |
| Full length recombinant protein of human TCP11L2 (NP_689985) produced in HEK293T cells. | |
| 0.1 mL | |
| 255394.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2A10 | |
| Flow Cytometry | |
| MGC40368, t-complex 11 (mouse) like 2, t-complex 11 (mouse)-like 2, T-complex protein 11-like protein 2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction