Learn More
TCP1 Mouse, anti-Human, CoraLite Plus 488, Clone: 2C2F6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68183-100UL
Description
The TCP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TCP1. This antibody recognizes human antigen. The TCP1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| TCP1 | |
| 2C2F6 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| CCT alpha, CCT1, CCTa, D6S230E, t complex 1, TCP 1 alpha, TCP1 | |
| 100 μL | |
| 6950 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |