Learn More
Invitrogen™ TCP1 Monoclonal Antibody (91a), PerCP

Rat Monoclonal Antibody
Brand: Invitrogen™ MA545720
Description
1 μg/mL of MA5-45720 was sufficient for detection of TCP1 alpha in 20 μg of 3T3 cell lysate by colorimetric immunoblot analysis using Goat anti-rat IgG:HRP as the secondary antibody. Detects approximately 60kDa. Also detects approximately 92kDa. Cross reactivity with human HSP60 has been observed with this antibody in immunoblot analysis. Reacts weakly with Saccharomyces cerevisiae, consistent with the epitope sequence being AKLRS (instead of AKLRA). In C. elegans, it reacts with TCP1 alpha and another CCT subunit. In plants, it recognizes TCP1 of Pisum sativum, and the sequence of Arabidopsis thalania TCP1 over the region of the epitope AKLRA. It has also been shown that it reacts with a subunit of a specialized chaperonin which folds phytochrome.
The T Complex Polypeptide 1 (TCP-1) is approximately 60 kDa protein constitutively expressed in almost all eukaryotic cells, and is upregulated during spermatogenesis. It is found in the cytosol as a subunit of a hetero-oligomeric chaperone that is known to be involved in the folding of actin and tubulin. The family of proteins termed chaperonins act to recognize and stabilize polypeptide intermediates during folding, assembly and disassembly, and share many characteristics with Heat Shock Protein 70 (HSP70) including high abundance, induction by environmental stress, and ATPase activity. The chaperonin family includes the mitochondrial HSP60, Escherichia coli GroEL, the plastid Rubisco-subunit binding protein, and the archaebacterial protein TF55. The TCP-1 sequence shows nearly 40% identity to TF55, but only minimal similarity to HSP60 and GroEL.
Specifications
| TCP1 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| P11983, P12612, P17987, P18279, P28480, Q32L40 | |
| TCP1 | |
| Recombinant Mouse TCP1 alpha protein fragment (carboxy terminal region). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Drosophila, Yeast, Yeast, C. elegans | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 91a | |
| PerCP | |
| TCP1 | |
| 65 kDa antigen; AI528772; c-cpn; CCT; Cct1; Ccta; CCTalpha; CCT-alpha; chaperonin-containing T-complex alpha subunit TCP1; chaperonin-containing T-complex polypeptide alpha subunit; cytosolic chaperonin containing t-complex polypeptide 1; D6S230E; hypothetical protein; p63; tailless complex polypeptide 1; tailless complex polypeptide 1A; Tailless complex polypeptide 1B; t-complex 1; t-complex polypeptide 1; t-complex protein 1; T-complex protein 1 alpha subunit-like protein; T-complex protein 1 subunit alpha; T-complex protein 1 subunit alpha A; T-complex protein 1 subunit alpha B; T-complex protein 1, alpha subunit; Tcp1; Tcp-1; TCP-1-A; TCP-1-alpha; TCP-1-B; Tp63; TRic; Unknown (protein for MGC:133746); YD8142.13; YD8142B.04; YDR212W | |
| Rat | |
| Protein G | |
| RUO | |
| 100153485, 100302435, 100689476, 100713821, 21454, 24818, 484064, 512043, 6950, 851798 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.