missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1-eta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35222-20ul
This item is not returnable.
View return policy
Description
TCP1-eta Polyclonal antibody specifically detects TCP1-eta in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TCP1-eta | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CCT-eta, CCTETA, CCTH, chaperonin containing TCP1, subunit 7 (eta), eta subunit, HIV-1 Nef interacting protein, HIV-1 Nef-interacting protein, MGC110985, Nip7-1, T-complex protein 1 subunit eta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 310-500 of human TCP1-eta (NP_006420.1).,, Sequence:, CAGRVPEEDLKRTMMACGGSIQTSVNALSADVLGRCQVFEETQIGGERYNFFTGCPKAKTCTFILRGGAEQFMEETERSLHDAIMIVRRAIKNDSVVAGGGAIEMELSKYLRDYSRTIPGKQQLLIGAYAKALEIIPRQLCDNAGFDATNILNKLRARHAQGGTWYGVDINNEDIADNFEAFVWEPAMVRI | |
| 20 μL | |
| Cell Cycle and Replication | |
| 10574 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction