missing translation for 'onlineSavingsMsg'
Learn More

TCP1-eta Antibody (1D6), Novus Biologicals™

Product Code. 18399298 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399298 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399298 Supplier Novus Biologicals Supplier No. H00010574M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TCP1-eta Monoclonal antibody specifically detects TCP1-eta in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP1-eta
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1D6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH19296
Gene Alias CCT-eta, CCTETA, CCTH, chaperonin containing TCP1, subunit 7 (eta), eta subunit, HIV-1 Nef interacting protein, HIV-1 Nef-interacting protein, MGC110985, Nip7-1, T-complex protein 1 subunit eta
Host Species Mouse
Immunogen CCT7 (AAH19296, 425 a.a. ~ 528 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RTIPGKQQLLIGAYAKALEIIPRQLCDNAGFDATNILNKLRARHAQGGTWYGVDINNEDIADNFEAFVWEPAMVRINALTAASEAACLIVSVDETIKNPRSTVD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10574
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.