missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1-delta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41361
This item is not returnable.
View return policy
Description
TCP1-delta Polyclonal antibody specifically detects TCP1-delta in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| TCP1-delta | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) | |
| CCTD, CCT-DELTA, chaperonin containing t-complex polypeptide 1, delta subunit, chaperonin containing TCP1, subunit 4 (delta), MGC126165, SRBMGC126164, Stimulator of TAR RNA-binding, T-complex protein 1 subunit delta, TCP-1-delta | |
| The epitope is recognized to a region between residue 489 to 539 of human Chaperonin Containing Tcp1, Subunit 4 (Delta). | |
| 100 μg | |
| Cell Cycle and Replication | |
| 10575 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000 - 1:5000, Immunoprecipitation 2 - 10 μg/mg lysate | |
| NP_006421.2 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction