missing translation for 'onlineSavingsMsg'
Learn More

TCP1-delta Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-41361CL1

Product Code. 30153886

  • £771.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TCP1-delta Polyclonal antibody specifically detects TCP1-delta in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TCP1-delta
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Immunoprecipitation, Western Blot
CoraFluor 1
CCTD, CCT-DELTA, chaperonin containing t-complex polypeptide 1, delta subunit, chaperonin containing TCP1, subunit 4 (delta), MGC126165, SRBMGC126164, Stimulator of TAR RNA-binding, T-complex protein 1 subunit delta, TCP-1-delta
The epitope is recognized to a region between residue 489 to 539 of human Chaperonin Containing Tcp1, Subunit 4 (Delta).
0.1 mL
Cell Cycle and Replication
10575
Store at 4°C in the dark. Do not freeze.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.