missing translation for 'onlineSavingsMsg'
Learn More

TCP1-beta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£288.00 - £451.00

Specifications

Antigen TCP1-beta
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18472331
View Documents
Novus Biologicals
NBP1-88146-25ul
25 μL
£288.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18242838
View Documents
Novus Biologicals
NBP1-88146
0.1 mL
£451.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TCP1-beta Polyclonal specifically detects TCP1-beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

TCP1-beta
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat
Cctb, CCT-beta, CCTBPRO1633,99D8.1, chaperonin containing t-complex polypeptide 1, beta subunit, chaperonin containing t-complex polypeptide 1, subunit 2, chaperonin containing TCP1, subunit 2 (beta), MGC142074, MGC142076, T-complex protein 1 subunit beta, T-complex protein 1, beta subunit, TCP-1-beta
CCT2
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500
Polyclonal
Rabbit
Cell Cycle and Replication, Membrane Trafficking and Chaperones
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
10576
This antibody was developed against Recombinant Protein corresponding to amino acids:VNQPKRIENAKILIANTGMDTDKIKIFGSRVRVDSTAKVAEIEHAEKEKMKEKVERILKHGINCFINRQLIYNYPEQLFGAAGVMAIEHADFAGVERLALVTGGEIASTFDHPELVKLGSCKLIEEVMIGEDKLIHFSGV
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.