missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1-beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88147-25ul
This item is not returnable.
View return policy
Description
TCP1-beta Polyclonal specifically detects TCP1-beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TCP1-beta | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Cctb, CCT-beta, CCTBPRO1633,99D8.1, chaperonin containing t-complex polypeptide 1, beta subunit, chaperonin containing t-complex polypeptide 1, subunit 2, chaperonin containing TCP1, subunit 2 (beta), MGC142074, MGC142076, T-complex protein 1 subunit beta, T-complex protein 1, beta subunit, TCP-1-beta | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CCT2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DALCVLAQTVKDSRTVYGGGCSEMLMAHAVTQLANRTPGKEAVAMESYAKALRMLPTIIADNAGYDSADLVAQLRAAHSEGNTTAGLDMREGTIGDMAILGITESFQVKRQVLLSAAEAAEVILRVDN | |
| 25 μL | |
| Cell Cycle and Replication, Membrane Trafficking and Chaperones | |
| 10576 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction