missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1 alpha Rabbit anti-Human, Clone: 3G9G8, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £395.00
Specifications
| Antigen | TCP1 alpha |
|---|---|
| Clone | 3G9G8 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18377576
|
Novus Biologicals
NBP3-16290-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18355515
|
Novus Biologicals
NBP3-16290-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCP1 alpha Monoclonal antibody specifically detects TCP1 alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TCP1 alpha | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TCP1 alpha (P17987). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 3G9G8 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6950 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title