missing translation for 'onlineSavingsMsg'
Learn More

TCP1 alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£292.00 - £435.00

Specifications

Antigen TCP1 alpha
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500-1:1000
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18493111
View Documents
Novus Biologicals
NBP1-88148-25ul
25 μL
£292.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18246386
View Documents
Novus Biologicals
NBP1-88148
0.1 mL
£435.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TCP1 alpha Polyclonal specifically detects TCP1 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TCP1 alpha
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
RUO
Human, Mouse, Rat
CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha
TCP1
IgG
Affinity Purified
Specificity of human TCP1 alpha antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500-1:1000
Polyclonal
Rabbit
Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
6950
This antibody was developed against Recombinant Protein corresponding to amino acids:CVVKRVLESKSVVPGGGAVEAALSIYLENYATSMGSREQLAIAEFARSLLVIPNTLAVNAAQDSTDLVAKLRAFHNEAQVNPERK
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.