missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00 - £434.00
Specifications
| Antigen | TCP1 alpha |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30226879
|
Novus Biologicals
NBP3-35903-20ul |
20 μL |
£154.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228604
|
Novus Biologicals
NBP3-35903-100ul |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCP1 alpha Polyclonal antibody specifically detects TCP1 alpha in Human,Rat samples. It is validated for ELISA,Western BlotSpecifications
| TCP1 alpha | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 6950 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 307-556 of human TCP1 alpha (NP_110379.2).,, Sequence:, AVRRVLKRDLKRIAKASGATILSTLANLEGEETFEAAMLGQAEEVVQERICDDELILIKNTKARTSASIILRGANDFMCDEMERSLHDALCVVKRVLESKSVVPGGGAVEAALSIYLENYATSMGSREQLAIAEFARSLLVIPNTLAVNAAQDSTDLVAKLRAFHNEAQVNPERKNLKWIGLDLSNGKPRDNKQAGVFEPTIVKVKSLKFATEAAITILRIDDLIKLHPESKDDKHGSYEDAVHSGALND | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title