missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP-1 zeta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561036
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RGLQDVLRTN LGPKGTMKML VSGAGDIKLT KDGNVL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 100%.
The protein TCP-1 (t complex polypeptide 1) is a subunit of the heterooligomeric complex CCT (chaperonin containing TCP-1) present in the eukaryotic cytosol. The CCT of eukaryotic cytosol is composed of eight different subunit species, TCP-1 alpha, beta, gamma, delta, epsilon, zeta, eta and theta, each encoded by a different gene. Two zeta subunits have been described: TCP-1 zeta (also designated TCP-1 zeta1) and TCP-1 zeta2. TCP-1 subunits are proposed to have independent functions in folding its in vivo substrates, the actins and tubulins. TCP-1 was first identified in the mouse as relevant for tail-less and embryonic lethal phenotypes. Sequences homologous to TCP-1 have been isolated in several other species, and the yeast TCP-1 has been shown to encode a molecular chaperone for Actin and Tubulin. TCP-1 found in mammalian cells and yeast plays an important role in the folding of cytosolic proteins.Specifications
TCP-1 zeta | |
Polyclonal | |
Unconjugated | |
CCT6A | |
acute morphine dependence related protein 2; acute morphine dependence-related protein 2; amino acid transport defect-complementing; CCT zeta1; CCT- zeta1; CCT- zeta-1; CCT zeta-1; Cct6; Cct6a; Cctz; Cctz1; Cctz-1; CCT-zeta; CCT-zeta-1; chaperonin containing T-complex subunit 6; chaperonin containing TCP-1; chaperonin containing TCP1 subunit 6A; chaperonin containing Tcp1, subunit 6A (zeta 1); chaperonin containing Tcp1, subunit 6a (zeta); chaperonin subunit 6a (zeta); histidine transp; histidine transport regulator 3; HTR3; MoDP-2; T-complex protein 1 subunit zeta; T-complex protein 1, zeta subunit; TCP 1 zeta; TCP1 zeta; TCP-1- zeta; TCP1- zeta; TCP-1-zeta; Tcp20; TCPZ; TTCP20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12466, 288620, 908 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P40227, P80317 | |
CCT6A | |
Recombinant protein corresponding to Human TCP-1 zeta | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only