missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCP-1 beta Recombinant Rabbit Monoclonal Antibody (24GB4170)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557005
This item is not returnable.
View return policy
Description
TCP-1 beta Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TCP-1 beta is a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). The CCT complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of the TCP-1 beta gene have been observed but not thoroughly characterized. A disease associated with TCP-1 beta protein dysfunction include epididymo-orchitis.
Specifications
| TCP-1 beta | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CCT2 | |
| 99D8.1; AI528772; c-cpn; CCT; Cct1; CCT2; Ccta; CCTB; CCT-beta; chaperonin containing t-complex polypeptide 1, beta subunit; chaperonin containing t-complex polypeptide 1, subunit 2; chaperonin containing TCP1 subunit 2; chaperonin containing Tcp1, subunit 2 (beta); chaperonin subunit 2 (beta); chaperonin-containing T-complex polypeptide beta subunit; epididymis secretory sperm binding protein Li 100n; HEL-S-100n; hypothetical protein LOC505313; p63; PRO1633; T-complex protein 1 subunit beta; T-complex protein 1, beta subunit; Tcp-1; TCP-1-beta; Tp63; TRic | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10576, 12461, 299809 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4170 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P78371, P80314, Q5XIM9 | |
| CCT2 | |
| A synthesized peptide derived from human TCP1 beta (aa 480-535). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction