missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCP-1 beta Monoclonal Antibody (5B5F5)

Product Code. 12807922
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12807922 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12807922 Supplier Invitrogen™ Supplier No. MA515901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-15901 targets CCT2 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15901 immunogen is purified recombinant fragment of human CCT2 expressed in E. Coli. . MA5-15901 detects CCT2 which has a predicted molecular weight of approximately 54kDa.

TCP-1 beta is a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). The CCT complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of the TCP-1 beta gene have been observed but not thoroughly characterized. A disease associated with TCP-1 beta protein dysfunction include epididymo-orchitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP-1 beta
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5B5F5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene CCT2
Gene Accession No. P78371, P80314, Q5XIM9
Gene Alias 99D8.1; AI528772; c-cpn; CCT; Cct1; CCT2; Ccta; CCTB; CCT-beta; chaperonin containing t-complex polypeptide 1, beta subunit; chaperonin containing t-complex polypeptide 1, subunit 2; chaperonin containing TCP1 subunit 2; chaperonin containing Tcp1, subunit 2 (beta); chaperonin subunit 2 (beta); chaperonin-containing T-complex polypeptide beta subunit; epididymis secretory sperm binding protein Li 100n; HEL-S-100n; hypothetical protein LOC505313; p63; PRO1633; T-complex protein 1 subunit beta; T-complex protein 1, beta subunit; Tcp-1; TCP-1-beta; Tp63; TRic
Gene Symbols CCT2
Host Species Mouse
Immunogen Purified recombinant fragment of human CCT2 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10576, 12461, 299809
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.