Learn More
Invitrogen™ TCOF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120816
Description
Positive test controls include: BT-474, 293T. The target is usually found in the following locations: Nucleus, nucleolus. Immunogen sequence: VRVGTQAPRK AGTATSPAGS SPAVAGGTQR PAEDSSSSEE SDSEEEKTGL AVTVGQAKSV GKGLQVKAAS VPVKGSLGQG TAPVLPGKTG PTVTQVKAEK QEDSESSEEE SDSEEAAASP AQVKTSVKKT QAKANPAAAR APSAKGTISA PGKVVTAAAQ AKQRSPSKVK PPVRNPQNST VLARGPASVP SVGKAVATAA QAQTGPEEDS GSSEEESDSE EEAETLAQVK PSGKTHQIRA ALAPAKESPR KGAAPTPPGK TGPSAAQAGK QDDSGSSSEE SDSDGEAPAA VTSAQDQESS.
Treacle is a serine/alanine-rich 1,411 amino acid nuclear phosphoprotein that interacts with upstream binding factor (UBF) and affects transcription of the ribosomal DNA gene. Treacle is also involved in early embryonic development, particularly in the craniofacial complex, and may play a role in nucleolarcytoplasmic transport. The Treacle protein contains three domains with unique N- and C-termini and a large central repeat domain. Mutations in TCOF1, the gene that encodes for Treacle, cause Treacher Collins-Franceschetti syndrome (TCS), a disorder characterized by defects in craniofacial development. Symptoms of TCS include conductive hearing loss, hypoplasia of the mandible and maxilla, downward sloping palpebral fissures and cleft palate.
Specifications
| TCOF1 | |
| Polyclonal | |
| Unconjugated | |
| TCOF1 | |
| AA408847; AW209012; MFD1; nucleolar trafficking phosphoprotein; Tcof1; TCS; TCS1; Treacher Collins Franceschetti syndrome 1, homolog; Treacher Collins syndrome protein; Treacher Collins syndrome protein homolog; Treacher Collins-Franceschetti syndrome 1; Treacher Collins-Franceschetti syndrome 1 homolog; treacle; Treacle protein; treacle ribosome biogenesis factor 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6949 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.91 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13428 | |
| TCOF1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 659-958 of human TCOF1 (NP_001008657.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.