missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCOF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575250
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Treacle is a serine/alanine-rich 1,411 amino acid nuclear phosphoprotein that interacts with upstream binding factor (UBF) and affects transcription of the ribosomal DNA gene. Treacle is also involved in early embryonic development, particularly in the craniofacial complex, and may play a role in nucleolarcytoplasmic transport. The Treacle protein contains three domains with unique N- and C-termini and a large central repeat domain. Mutations in TCOF1, the gene that encodes for Treacle, cause Treacher Collins-Franceschetti syndrome (TCS), a disorder characterized by defects in craniofacial development. Symptoms of TCS include conductive hearing loss, hypoplasia of the mandible and maxilla, downward sloping palpebral fissures and cleft palate.
Specifications
| TCOF1 | |
| Polyclonal | |
| Unconjugated | |
| TCOF1 | |
| AA408847; AW209012; MFD1; nucleolar trafficking phosphoprotein; Tcof1; TCS; TCS1; Treacher Collins Franceschetti syndrome 1, homolog; Treacher Collins syndrome protein; Treacher Collins syndrome protein homolog; Treacher Collins-Franceschetti syndrome 1; Treacher Collins-Franceschetti syndrome 1 homolog; treacle; Treacle protein; treacle ribosome biogenesis factor 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21453, 291571, 6949 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O08784, Q13428 | |
| TCOF1 | |
| Synthetic peptide corresponding to amino acids 22-73 of Human TCOF1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction