missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCN238, MedChemExpress
Description
TCN238 is an orally bioavailable mGlu4 receptor positive allosteric modulator (PAM) with an EC50 of 1 μM.
Specifications
Specifications
| Chemical Name or Material | TCN238 |
| Physical Form | Solid |
| CAS | 125404-04-8 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C12H11N3 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : ≥ 150 mg/mL (760.49 mM) |
| SMILES | NC1=NC=CC(/C=C/C2=CC=CC=C2)=N1 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?