missing translation for 'onlineSavingsMsg'
Learn More

TCN1 Antibody (3F10), Novus Biologicals™

Product Code. 18361409 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361409 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361409 Supplier Novus Biologicals Supplier No. H00006947M12

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TCN1 Monoclonal antibody specifically detects TCN1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCN1
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3F10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Alias haptocorin, TC I, TC-1, TC1haptocorrin, TCItranscobalamin-1, Transcobalamin I, transcobalamin I (vitamin B12 binding protein, R binder family)
Host Species Mouse
Immunogen TCN1 (AAH18632, 1 a.a. ~ 423 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRQSHQLPLVGLLLFSFIPSQLCEICEVSEENYIRLKPLLNTMIQSNYNRGTSAVNVVLSLKLVGIQIQTLMQKMIQQIKYNVKSRLSDVSSGELALIILALGVCRNAEENLIYDYHLIDKLENKFQAEIENMEAHNGTPLTNYYQLSLDVLALCLFNGNYSTAEVVNHFTPENKNYYFGSQFSVDTGAMAVLALTCVKKSLINGQIKADEGSLKNISIYTKSLVEKILSEKKENGLIGNTFSTGEAMQALFVSSDYYNENDWNCRQTLNTVLTEISQGAFSNPNAAAQVLPALMGKTFLDINKDSSCVPASGNFNISADEPITVTPPDSQSYISVDYSVRINETYFTNVAVLNGSVFLSVMEKAQKMNDTIFGFTMEERSWGPYITCIQGLCANNNDRTYWELLSGGEPLSQGAGSYVVRNG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6947
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.