missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TCN 201

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
NMDA receptor antagonist selective for GluN1/GluN2A (formally NR1/NR2A) over GluN1/GluN2B (formally NR1/NR2B) containing receptors (pIC50 values are 6.8 and <4.3, respectively, in human recombinant GluN1/GluN2A (formally NR1/NR2A) and GluN1/GluN2B (formally NR1/NR2B) FLIPR/Ca2+ assays).
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 461.89 |
| Purity | >99% |
| Molecular Weight (g/mol) | 461.892 |
| Chemical Name or Material | TCN 201 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction