missing translation for 'onlineSavingsMsg'
Learn More

TCLP Volatiles Spike, SPEX CertiPrep™

Product Code. 16330498 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16330498 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16330498 Supplier SPEX Certiprep Supplier No. TCLPVX
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,1-Dichloroethene75-35-42000μg/mL
1,2-Dichloroethane107-06-22000μg/mL
1,4-Dichlorobenzene106-46-72000μg/mL
2-Butanone78-93-32000μg/mL
Benzene71-43-22000μg/mL
Carbon tetrachloride56-23-52000μg/mL
Chlorobenzene108-90-72000μg/mL
Chloroform67-66-32000μg/mL
Tetrachloroethene127-18-42000μg/mL
Trichloroethene79-01-62000μg/mL
Vinyl Chloride75-01-42000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Volatiles Spike
Name Note USEPA Methods 1311, 1312
Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Methods For US EPA Methods 1311, 1312, 8260B
Technique AA, ICP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methanol-P and T
Quantity 1 mL

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.