missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCL1A Rabbit anti-Human, Clone: TCL1/2747R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | TCL1A |
|---|---|
| Clone | TCL1/2747R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
TCL1A Monoclonal specifically detects TCL1A in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| TCL1A | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 8115 | |
| IgG | |
| Protein A or G purified |
| TCL1/2747R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A | |
| Recombinant human TCL1A protein fragment (around aa 2-109) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title